MilliporeSigma
MIPUP: minimum perfect unmixed phylogenies for multi-sampled tumors via branchings and ILP.
Bioinformatics (Oxford, England) (2018-08-14)
Edin Husić, Xinyue Li, Ademir Hujdurović, Miika Mehine, Romeo Rizzi, Veli Mäkinen, Martin Milanič, Alexandru I Tomescu
RÉSUMÉ
Discovering the evolution of a tumor may help identify driver mutations and provide a more comprehensive view on the history of the tumor. Recent studies have tackled this problem using multiple samples sequenced from a tumor, and due to clinical implications, this has attracted great interest. However, such samples usually mix several distinct tumor subclones, which confounds the discovery of the tumor phylogeny. We study a natural problem formulation requiring to decompose the tumor samples into several subclones with the objective of forming a minimum perfect phylogeny. We propose an Integer Linear Programming formulation for it, and implement it into a method called MIPUP. We tested the ability of MIPUP and of four popular tools LICHeE, AncesTree, CITUP, Treeomics to reconstruct the tumor phylogeny. On simulated data, MIPUP shows up to a 34% improvement under the ancestor-descendant relations metric. On four real datasets, MIPUP's reconstructions proved to be generally more faithful than those of LICHeE. MIPUP is available at https://github.com/zhero9/MIPUP as open source. Supplementary data are available at Bioinformatics online.