MilliporeSigma
Use of Physcion to Improve Atopic Dermatitis-Like Skin Lesions through Blocking of Thymic Stromal Lymphopoietin.
Molecules (Basel, Switzerland) (2019-04-18)
Phil-Dong Moon, Na-Ra Han, Jin Soo Lee, Sungwei Hong, Min-Sun Yoo, Hyeong-Jin Kim, Ji-Hyeon Kim, Soonsik Kang, Hyun-Woo Jee, Hyung-Min Kim, Hyun-Ja Jeong
RÉSUMÉ
Physcion is well known for the treatment of carcinoma. However, the therapeutic effect of physcion on atopic dermatitis (AD) through the inhibition of thymic stromal lymphopoietin (TSLP) level remains largely unknown. In this study, we investigated the anti-AD effect of physcion using HMC-1 cells, splenocytes, and a murine model. Treatment with physcion decreased production and mRNA expression levels of TSLP, IL-6, TNF-ɑ, and IL-1β in activated HMC-1 cells. Physcion reduced the expression levels of RIP2/caspase-1 and phospho (p)ERK/pJNK/pp38 in activated HMC-1 cells. Physcion suppressed the expression levels of pIKKβ/NF-κB/pIkB in activated HMC-1 cells. Moreover, physcion attenuated the production levels of TSLP, IL-4, IL-6, TNF-, and IFN-γ from activated splenocytes. Oral administration of physcion improved the severity of 2,4-dinitrochlorobenzene-induced AD-like lesional skin through reducing infiltration of inflammatory cells and mast cells, and the protein and mRNA levels of TSLP, IL-4, and IL-6 in the lesional skin tissues. Physcion attenuated histamine, IgE, TSLP, IL-4, IL-6, and TNF- levels in serum. In addition, physcion inhibited caspase-1 activation in the lesional skin tissues. These findings indicate that physcion could ameliorate AD-like skin lesions by inhibiting TSLP levels via caspase-1/MAPKs/NF-kB signalings, which would provide experimental evidence of the therapeutic potential of physcion for AD.