MilliporeSigma
Optically Active Hybrid Materials Based on Natural Spider Silk.
ACS applied materials & interfaces (2019-06-30)
Aleksandra Kiseleva, Grigorii Kiselev, Vadim Kessler, Gulaim Seisenbaeva, Dmitry Gets, Valeriya Rumyantseva, Tatiana Lyalina, Anna Fakhardo, Pavel Krivoshapkin, Elena Krivoshapkina
RÉSUMÉ
Spider silk is a natural material possessing unique properties such as biocompatibility, regenerative and antimicrobial activity, and biodegradability. It is broadly considered an attractive matrix for tissue regeneration applications. Optical monitoring and potential control over tissue regrowth are attractive tools for monitoring of this process. In this work, we show upconversion modification of natural spider silk fibers with inorganic nanoparticles. To achieve upconversion, metal oxide nanoparticles were doped with low concentrations of rare-earth elements, producing potentially biocompatible luminescent nanomaterials. The suggested approach to spider silk modification is efficient and easy to perform, opening up sensing and imaging possibilities of biomaterials in a noninvasive and real-time manner in bio-integration approaches.