MilliporeSigma
Lipid droplets participate in modulating innate immune genes in Ctenopharyngodon idella kidney cells.
Fish & shellfish immunology (2019-03-21)
Cai-Xia Lei, Jing-Jing Tian, Wen Zhang, Yu-Ping Li, Hong Ji, Er-Meng Yu, Wang-Bao Gong, Zhi-Fei Li, Kai Zhang, Guang-Jun Wang, De-Guang Yu, Jun Xie
RÉSUMÉ
Lipid droplets (LDs) are increasingly being recognized as important immune modulators in mammals, in additional to their function of lipid ester deposition. However, the role of LDs in fish immunity remains poorly understood. In this study, the function of LDs in the innate immune response of Ctenopharyngodon idella kidney (CIK) cells, which are the equivalent of myeloid cells in vertebrates, was investigated. LD number and TG content significantly increased in the CIK cells following exposure to lipopolysaccharide (LPS), peptidoglycan (PGN), and polyriboinosinic-polyribocytidylic acid (Poly [I: C]) for 24 h, accompanied by increases in the relative expression of several innate immune genes. However, fatty acid compositions of the triglycerides were not changed after treatment with these three pathogenic mimics. LPS, PGN, and Poly (I: C) did not alter the relative expressions of lipogenic (FAS, SCD, and DGAT) and lipid catabolic (PPARα, ATGL, and CPT-1) genes. However, these treatments did increase the mRNA levels of lipid transportation genes (FATP/CD36, ACSL1, and ACSL4), and also decreased the non-esterified fatty acid level in the medium. To further explore the role of LDs in the immune response, CIK cells were incubated with different concentrations (0, 100, 200, 300, 400, 500 μM) of exogenous lipid mix (LM; oleic acid [OA]:linoleic acid [LA]:linolenic acid [LNA] = 2:1:1), and were then transferred to a lipid-free medium and incubated for 24 h. LD size and number increased with the increase in lipid levels, and this was accompanied by increased expression of innate immune genes, including MyD88, IRF3, and IL-1β, which were expressed at their highest levels in 300 μM exogenous lipid mix. Interestingly, after incubating with different fatty acids (LM, OA, LA, LNA, arachidonic acid [ARA], and docosahexaenoic acid [DHA]; 300 μM), ARA and DHA were more potent in inducing LD formation and innate immune gene expression in the CIK cells. Finally, atglistatin, an ATGL inhibitor, effectively attenuated the expression of most genes upregulated by ARA or DHA, suggesting that lipolysis may be involved in the regulation of immune genes at the transcriptional level. Overall, the findings of this study demonstrate that LDs are functional organelles that could act as modulators in the innate immune response of CIK cells. Additionally, long-chain polyunsaturated fatty acid enriched LDs play a unique role in regulating this process.