MilliporeSigma
  • Development and validation of probe drug cocktails for the characterization of CYP450-mediated metabolism by human heart microsomes.
Development and validation of probe drug cocktails for the characterization of CYP450-mediated metabolism by human heart microsomes.
Xenobiotica; the fate of foreign compounds in biological systems (2018-02-17)
Jade Huguet, Fleur Gaudette, Veronique Michaud, Jacques Turgeon
RÉSUMÉ
1. The objective of our study was to develop and validate a cocktail approach to allow the simultaneous characterization of various CYP450-mediated oxidations by human heart microsomes for nine probe drug substrates, namely, 7-ethoxyresorufin, bupropion, repaglinide, tolbutamide, bufuralol, chlorzoxazone, ebastine, midazolam and dodecanoic acid. 2. The first validation step was conducted using recombinant human CYP450 isoenzymes by comparing activity measured for each probe drug as a function of (1) buffer used, (2) selectivity towards specific isoenzymes and (3) drug interactions between probes. Activity was all measured by validated LC-MSMS methods. 3. Two cocktails were then constituted with seven of the nine drugs and subjected to kinetic validation. Finally, all probe drugs were incubated with human heart microsomes prepared from ventricular tissues obtained from 12 patients undergoing cardiac transplantation. 4. Validated cocktail #1 including bupropion, chlorzoxazone, ebastine and midazolam was used to characterize CYP2B6-, 2E1-, 2J2- and 3A5-mediated metabolism in human hearts. 5. Cocktail #2 which includes bufuralol, 7-ethoxyresorufin and repaglinide failed the validation step. Substrates in cocktail #2 as well as tolbutamide and dodecanoic acid had to be incubated separately because of their physico-chemical characteristics (solubility and ionization) or drug interactions. 6. Activity in HHM was the highest towards ebastine, chlorzoxazone and tolbutamide.