MilliporeSigma
Apigenin attenuates doxorubicin induced cardiotoxicity via reducing oxidative stress and apoptosis in male rats.
Life sciences (2019-07-08)
Masoud Fallah Rajabpour Zare, Kamran Rakhshan, Nahid Aboutaleb, Farnaz Nikbakht, Nasim Naderi, Morteza Bakhshesh, Yaser Azizi
RÉSUMÉ
Doxorubicin, an antibiotic belonging to anthracycline family, has been used for treatment of malignancies. Cardiotoxicity is the main adverse effect of doxorubicin. Apigenin, as a flavonoid, has antioxidant, anti-inflammatory and anti-tumoral properties. The aim of this study was the assessment of any protective effect of apigenin on cardiotoxicity induced by doxorubicin. 40 male Wistar rats were randomly divided into 4 groups: control, cardiotoxicity (DOX), apigenin treated group (DOX + Api 25) and apigenin group (Api 25). At the end of the experiment, the markers of cardiac function (%EF, %FS, LVIDs, LVIDd), cardiac and liver injury (LDH, CK-MB, cTn-I, ALT, and AST), cardiac apoptosis (Bax, Bcl-2 and Caspase3), cardiac oxidative stress (SOD, GSH, MDA) and cardiac fibrosis were measured. Apigenin improved cardiac functional parameters. The levels of cardiac and liver injury markers were significantly decreased in DOX + Api 25 compared to DOX. Treatment with apigenin caused significant decrease in percentage of cardiac fibrosis in comparison with DOX. Apigenin in DOX + Api 25 group led to significant decrease in apoptotic proteins (Casp3, Bax) and a significant increase in anti-apoptotic proteins (Bcl2). In apigenin treatment groups, SOD levels significantly increased while a significant decrease was observed in MDA. The amount of GSH in DOX + Api 25 had no significant change in comparison to control and Api 25 groups. Apigenin reduced cardiac injuries induced by DOX through anti-fibrotic, antioxidant and anti-apoptotic properties. It seems that apigenin prevents cardiac injuries and improves cardiac function.