MilliporeSigma
  • Insights into the Regulation Effects of Certain Phenolic Acids on 2,3-Dihydro-3,5-dihydroxy-6-methyl-4(H)-pyran-4-one Formation in a Microaqueous Glucose-Proline System.
Insights into the Regulation Effects of Certain Phenolic Acids on 2,3-Dihydro-3,5-dihydroxy-6-methyl-4(H)-pyran-4-one Formation in a Microaqueous Glucose-Proline System.
Journal of agricultural and food chemistry (2019-07-25)
He Li, Chun-Jian Wu, Xiang-Yi Tang, Shu-Juan Yu
RÉSUMÉ
The control of 2,3-dihydro-3,5-dihydroxy-6-methyl-4(H)-pyran-4-one (DDMP) formation in the Maillard reaction is important to improve the thermally treated food quality as a result of its intense bitterness and potential toxicity. In this work, phenolic acids, such as gallic, protocatechuic, caffeic, and ferulic acids, were applied to modulate DDMP formation in a microaqueous glucose-proline model. The formation of DDMP was inhibited at low concentrations (from 0.1 to 5.0 mM) while enhanced at 10.0 mM gallic, protocatechuic, and caffeic acids. Ferulic acid always inhibited DDMP formation as a result of the absence of catechol groups on its benzene ring. The result indicated that the control of DDMP formation depended upon the concentration and chemical structures of phenolic acids, such as the number of hydroxyl groups. Further studies indicated that the hydroxyl distribution of phenolic acids regulated the peroxide formation in the model reaction system and further changed the development of the oxidation reaction, which affected the degradation of glucose via caramel or Maillard reaction, Amadori rearrangement product oxidation, and 1-deoxyglucosone degradation to form the intermediates.