MilliporeSigma
Impact of Fractional Ablative Carbon Dioxide Laser on the Treatment of Rhinophyma.
Aesthetic surgery journal (2018-09-12)
Maine Trece de Siqueira Santos do Amaral, Alessandra Haddad, Fabio Xerfan Nahas, Yara Juliano, Lydia Masako Ferreira
RÉSUMÉ
Rhinophyma results in nasal deformity, which may lead to significant distress, embarrassment, social isolation, and low self-esteem. Fractional carbon dioxide (CO2) laser treatment is a simple, noninvasive, outpatient procedure with a low complication rate that has proved useful in rhinophyma treatment. The aim of this study was to assess the impact of fractional ablative CO2 laser treatment on self-esteem in patients with rhinophyma. Twelve patients with rhinophyma, between 50 and 80 years of age, received 4 monthly treatment sessions with a fractional ablative CO2 laser. The laser parameters were chosen based on the severity of the condition of each patient. Patient satisfaction was assessed through a study-specific questionnaire administered 1 month after treatment. Self-esteem was assessed by the Rosenberg Self-Esteem (RSE) Scale, which was administered before treatment, and at 1, 6, and 12 months after treatment. A decrease in RSE scores was observed in the post-treatment period, indicating an improvement in self-esteem among patients. All patients were satisfied with the aesthetic results and would recommend the treatment to a friend. Fractional ablative CO2 laser treatment is associated with high levels of rhinophyma patient satisfaction, and significant improvements in self-esteem.