MilliporeSigma
  • Inflammatory and regulatory CCL and CXCL chemokine and cytokine cellular responses in patients with patent Mansonella perstans filariasis.
Inflammatory and regulatory CCL and CXCL chemokine and cytokine cellular responses in patients with patent Mansonella perstans filariasis.
Clinical and experimental immunology (2018-12-19)
B Wangala, R G Gantin, P S Voßberg, A Vovor, W P Poutouli, K Komlan, M Banla, C Köhler, P T Soboslay
RÉSUMÉ
Mansonella perstans (Mp) filariasis is present in large populations in sub-Saharan Africa, and to what extent patent Mp infection modulates the expression of immunity in patients, notably their cellular cytokine and chemokine response profile, remains not well known. We studied the spontaneous and inducible cellular production of chemokines (C-X-C motif) ligand 9 (CXCL9) [monokine induced by interferon (IFN)-γ (MIG)], CXCL-10 [inducible protein (IP)-10], chemokine (C-C motif) ligand 24 (CCL24) (eotaxin-2), CCL22 [macrophage-derived chemokine (MDC)], CCL13 [monocyte chemotactic protein-4 (MCP-4)], CCL18 [pulmonary and activation-regulated chemokine (PARC)], CCL17 [thymus- and activation-regulated chemokine (TARC)] and interleukin (IL)-27 in mansonelliasis patients (Mp-PAT) and mansonelliasis-free controls (CTRL). Freshly isolated peripheral mononuclear blood cells (PBMC) were stimulated with helminth, protozoan and bacterial antigens and mitogen [phytohaemagglutinin (PHA)]. PBMC from Mp-PAT produced spontaneously (without antigen stimulation) significantly higher levels of eotaxin-2, IL-27, IL-8, MCP-4 and MDC than cells from CTRL, while IFN-γ-IP-10 was lower in Mp-PAT. Helminth antigens activated IL-27 and MCP-4 only in CTRL, while Ascaris antigen, Onchocerca antigen, Schistosoma antigen, Entamoeba antigen, Streptococcus antigen, Mycobacteria antigen and PHA stimulated MIG release in CTRL and Mp-PAT. Notably, Entamoeba antigen and PHA strongly depressed (P < 0·0001) eotaxin-2 (CCL24) production in both study groups. Multiple regression analyses disclosed in Mp-PAT and CTRL dissimilar cellular chemokine and cytokine production levels being higher in Mp-PAT for CCL24, IL-27, IL-8, MCP-4, MDC and PARC (for all P < 0·0001), at baseline (P < 0·0001), in response to Entamoeba histolytica strain HM1 antigen (EhAg) (P < 0·0001), Onchocerca volvulus adult worm-derived antigen (OvAg) (P = 0·005), PHA (P < 0·0001) and purified protein derivative (PPD) (P < 0·0001) stimulation. In Mp-PAT with hookworm co-infection, the cellular chemokine production of CXCL10 (IP-10) was diminished. In summary, the chemokine and cytokine responses in Mp-PAT were in general not depressed, PBMC from Mp-PAT produced spontaneously and selectively inducible inflammatory and regulatory chemokines and cytokines at higher levels than CTRL and such diverse and distinctive reactivity supports that patent M. perstans infection will not polarize innate and adaptive cellular immune responsiveness in patients.