MilliporeSigma
Sonic hedgehog pathway activation regulates cervical cancer stem cell characteristics during epithelial to mesenchymal transition.
Journal of cellular physiology (2019-02-05)
Anuka Sharma, Renaissa De, Shifa Javed, Radhika Srinivasan, Arnab Pal, Shalmoli Bhattacharyya
RÉSUMÉ
Resistance to therapy and metastasis remains one of the leading causes of mortality due to cervical cancer despite advances in detection and treatment. The mechanism of epithelial to mesenchymal transition (EMT) provides conceptual explanation to the invasiveness and metastatic spread of cancer but it has not been fully understood in cervical cancer. This study aims to investigate the mechanism by which silencing of E-cadherin gene regulates EMT leading to proliferation, invasion, and chemoresistance of cervical cancer cells through the Hedgehog (Hh) signaling pathway. We developed an in vitro EMT model by the knockdown of E-cadherin expression in cervical cancer cell lines. To understand the role of developmental pathway like Hh in the progression of cervical cancer, we investigated the expression of Hh pathway mediators by array in E-cadherin low cervical cancer cells and observed upregulation of Hh pathway. This was further validated on low passage patient-derived cell lines and cervical carcinoma tissue sections from cervical cancer patients. Further, we evaluated the role of two inhibitors (cyclopamine and GANT58) of the Hh pathway on invasiveness and apoptosis in E-cadherin low cervical cancer cells. In conclusion, we observed that inhibition of Hh pathway with GANT58 along with current therapeutic procedures could be more effective in targeting drug-resistant EMT cells and bulk tumor cells in cervical cancer.