MilliporeSigma
Identification of small molecule flavor compounds that contribute to the somatosensory attributes of bovine milk products.
Food chemistry (2019-05-28)
David M Potts, Devin G Peterson
RÉSUMÉ
Compounds that contribute to the somatosensory flavor profile of bovine fluid milk products were investigated. Sensory descriptive analysis defined five main attributes that consisted of "mouthcoating, astringent/drying, fatty texture, dairy mouthfeel, and tingling/irritation" sensations. Utilizing multi-dimensional LC sensory guided fractionation, compounds with these attributes were selected, purified and subsequently identified by LC/MS as orotic acid, pantothenic acid, hippuric acid, and p-cresol sulfate. Quantitative analysis of the four compounds across skim milk, low fat milk and whole milk indicated the concentrations were not significantly different; however, they were significantly lower in cream. Sensory recombination milk model analysis of each compound at endogenous concentrations of fluid milk indicated all compounds were sensory active. Furthermore, using a 2-AFC sensory test, skim milk spiked at two-fold higher concentrations of the 4 compounds had a significantly "creamier, fuller body" when compared with skim milk itself (α = 0.01).