MilliporeSigma
  • Inhibitory effects of antihypertensive drugs on human cytochrome P450 2J2 activity: Potent inhibition by azelnidipine and manidipine.
Inhibitory effects of antihypertensive drugs on human cytochrome P450 2J2 activity: Potent inhibition by azelnidipine and manidipine.
Chemico-biological interactions (2019-04-10)
Noriaki Ikemura, Satoshi Yamaori, Chinatsu Kobayashi, Shinobu Kamijo, Norie Murayama, Hiroshi Yamazaki, Shigeru Ohmori
RÉSUMÉ
The inhibitory effects of antihypertensive drugs (dihydropyridine calcium channel blockers, angiotensin II receptor blockers, and angiotensin-converting enzyme inhibitors) on cytochrome P450 2J2 (CYP2J2) activity were examined. Amlodipine, azelnidipine, barnidipine, benidipine, cilnidipine, efonidipine, felodipine, manidipine, nicardipine, nifedipine, nilvadipine, nisoldipine, nitrendipine, telmisartan, delapril, and quinapril inhibited luciferin-2J2/4F12 O-dealkylase activity of recombinant human CYP2J2 in a concentration-dependent manner (IC50 = 0.116-9.19 μM). Kinetic analyses of the inhibition indicated that azelnidipine, barnidipine, benidipine, cilnidipine, efonidipine, manidipine, nicardipine, telmisartan, delapril, and quinapril competitively inhibited CYP2J2 activity, while amlodipine, felodipine, nifedipine, nilvadipine, nisoldipine, and nitrendipine showed mixed inhibition. Among these drugs, manidipine showed the strongest reversible inhibition with Ki value of 0.0294 μM. The docking simulation data supported the potent inhibition of CYP2J2 by these drugs. Next, the effect of preincubation on CYP2J2 inhibition was investigated to determine whether these antihypertensive drugs inhibited CYP2J2 activity in a metabolism-dependent manner. A 20-min preincubation of azelnidipine and felodipine in the presence of NADPH potentiated the inhibition of CYP2J2. Furthermore, kinetic analysis of the inactivation showed that azelnidipine caused a preincubation time- and concentration-dependent decrease in CYP2J2 activity yielding kinact/KI value of 105 l/mmol/min, although felodipine showed no preincubation time-dependent inhibition. The azelnidipine-mediated inactivation required NADPH. These results indicated that manidipine is a potent competitive reversible inhibitor while azelnidipine is a potent mechanism-based inactivator of human CYP2J2.