MilliporeSigma
  • Updates on the surface antigens of basophils: CD16 on basophils of patients with respiratory or insect venom allergy and the rejection of CD203c and CD63 externalization decoupling by bisindolylmaleimides.
Updates on the surface antigens of basophils: CD16 on basophils of patients with respiratory or insect venom allergy and the rejection of CD203c and CD63 externalization decoupling by bisindolylmaleimides.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology (2018-10-06)
Petr Heneberg, Kamila Riegerová, Adéla Říhová, Daniela Šimčíková, Petr Kučera
RÉSUMÉ
CD16 was previously suggested to be a new marker of basophils that is subject to downregulation by FcεRI crosslinking. Certain compounds, including supraoptimal concentrations of the PKC inhibitors, bisindolylmaleimides, decouple the release of granules containing CD203c, CD63 and histamine, and may thus help to identify the mechanisms related to the CD16 externalization. We hypothesized that CD16 is differentially expressed on the surface of basophils in patients with birch pollen or insect venom allergy and is subject to a regulation in response to allergens. We also employed CD203c and CD63 externalization decoupling by bisindolylmaleimides. We performed a basophil activation test coupled with CD16 and histamine detection using cells isolated from patients with allergy to birch pollen or insect venom and negative controls. We employed two PKC inhibitors, bisindolylmaleimide II and Ro 31-8220 at their supraoptimal concentrations and, after difficulties reproducing previously published data, we analyzed the fluorescence of these inhibitors alone. We identified the CD16 isoforms by sequencing nested RT-PCR amplicons from flow cytometry sorted basophils and by cleaving the CD16b GPI anchor using a phospholipase C. We provide the first evidence that CD16a is expressed as a surface antigen on a small subpopulation of human basophils in patients with respiratory and insect venom allergy, and this antigen shows increased surface expression following allergen challenge or FcεRI crosslinking. We rejected the apparent decoupling of the surface expression of basophil activation markers following the administration of bisindolylmaleimides. The inclusion of αCD16 in negative selection cocktails selects against a subset of basophils that are CD16+ or CD16dim . Using CD16dim basophils and unstained leucocytes, we show that previous studies with supraoptimal concentrations of bisindolylmaleimides are likely flawed and are not associated with the differential expression of CD203c and CD63.