MilliporeSigma
  • A Simple Colorimetric and Fluorescent Sensor to Detect Organophosphate Pesticides Based on Adenosine Triphosphate-Modified Gold Nanoparticles.
A Simple Colorimetric and Fluorescent Sensor to Detect Organophosphate Pesticides Based on Adenosine Triphosphate-Modified Gold Nanoparticles.
Sensors (Basel, Switzerland) (2018-12-20)
Xiaoxia Li, Haixin Cui, Zhanghua Zeng
RÉSUMÉ
A simple and dual modal (colorimetric and fluorescent) sensor for organophosphate pesticides with high sensitivity and selectivity using adenosine triphosphate (ATP)- and rhodamine B-modified gold nanoparticles (RB-AuNPs), was successfully fabricated. This detection for ethoprophos afforded colorimetric and fluorescence imaging changes visualization. The quantitative determination was linearly proportional to the amounts of ethoprophos in the range of a micromolar scale (4.0⁻15.0 µM). The limit of detection for ethoprophos was as low as 37.0 nM at 3σ/k. Moreover, the extent application of this simple assay was successfully demonstrated in tap water samples with high reliability and applicability, indicating remarkable application in real samples.