MilliporeSigma
TXNIP mediates high glucose-induced mitophagic flux and lysosome enlargement in human retinal pigment epithelial cells.
Biology open (2019-04-27)
Takhellambam S Devi, Thangal Yumnamcha, Fayi Yao, Mallika Somayajulu, Renu A Kowluru, Lalit P Singh
RÉSUMÉ
Thioredoxin-interacting protein (TXNIP) plays a critical role in oxidative stress, inflammation, apoptosis and the pathogenesis of diabetic retinopathy (DR). However, the role of TXNIP in high glucose-induced retinal pigment epithelium (RPE) dysfunction is still unknown. Here, we show that high glucose (HG; 25 mM,) significantly increases TXNIP expression at both the mRNA and protein levels when compared to low glucose (LG; 5.5 mM) in a human RPE cell line (ARPE-19) and primary human RPE (HRPE) cells. TXNIP upregulation is associated with mitochondrial membrane depolarization, fragmentation and mitophagic flux to lysosomes. We used confocal live-cell imaging of RPE cells expressing mt-Keima, a coral protein that emits green light in mitochondria (alkaline or neutral pH) and red light in the acidic lysosome, to measure mitophagic flux. We observed an elongated mitochondrial network of green mt-Keima under LG, which is fragmented in HG. Red mt-Keima accumulates in lysosomes as small punctate aggregations under LG in both ARPE-19 and HRPE cells, whereas they are significantly enlarged (two- to threefold) under HG. Lysosomal enlargement under HG is further illustrated by lysosomal membrane protein LAMP1-mCherry expression in both ARPE-19 and HRPE cells. Furthermore, HG causes lysosomal cathepsin L inactivation and pro-inflammatory caspase-1 activation in ARPE-19 cells. TXNIP knockdown by shRNA prevents mitochondrial fragmentation, mitophagic flux and lysosome enlargement under HG. In addition, antioxidant N-acetylcysteine (NAC) and Amlexanox (Amlx), an inhibitor of protein kinase TBK1 and of the mitophagic adaptors Optineurin (Optn) and Sequestosome 1 (p62/SQSTM1), prevent mitophagic flux and lysosome enlargement. These results suggest that TXNIP mediates several deleterious effects of high glucose on RPE, which may be implicated in the development of DR.