- Atmospheric Cold Plasma and Peracetic Acid-Based Hurdle Intervention To Reduce Salmonella on Raw Poultry Meat.
Atmospheric Cold Plasma and Peracetic Acid-Based Hurdle Intervention To Reduce Salmonella on Raw Poultry Meat.
Journal of food protection (2019-04-25)
Shreyak Chaplot, Barun Yadav, Byeonghwa Jeon, M S Roopesh
PMID31017813
RÉSUMÉ
Atmospheric cold plasma and peracetic acid-based hurdle approach for safety of poultry products was evaluated. Study demonstrates a significant synergetic approach to reducing Salmonella on raw poultry. Hurdle approach shows promising bacterial reduction but requires further optimization.