MilliporeSigma
BAI1‑associated protein 2‑like 2 is a potential biomarker in lung cancer.
Oncology reports (2018-11-30)
Lei Xu, Hua Du, Qiang Zhang, Chao Wang, Li Yan, Guoying Tian, Xiuhua Fu
RÉSUMÉ
Lung cancer is the leading cause of cancer‑related death worldwide. The underlying molecular mechanisms that trigger this disease remain largely unknown. The I‑BAR family is involved in regulating cell membrane formation and some members, such as BAIAP2L1, IRSp53 and MIM have been shown to participate in tumorigenic progression. However, the role of BAI1‑associated protein 2‑like 2 (BAIAP2L2) in cancer development is unclear. In the present study, we determined that BAIAP2L2 was upregulated in lung adenocarcinoma tissues and various lung cancer cell lines. In vitro, BAIAP2L2 silencing resulted in decreased viability and colony formation capacity of both A549 and H1299 cells. By contrast, BAIAP2L2 overexpression promoted the proliferation and growth of 95D cells. These results indicated that BAIAP2L2 was essential for lung cancer cell proliferation and growth. We also found that BAIAP2L2 knockdown increased the apoptosis of A549 and H1299 cells. At the molecular level, BAIAP2L2 knockdown led to dysregulation of numerous genes, among which the Estrogen‑mediated S‑phase Entry pathway was significantly suppressed. Collectively, our findings revealed BAIAP2L2 as a novel biomarker and potential therapeutic target for lung cancer.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-BAIAP2L2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution