MilliporeSigma
  • Knockdown of TRPV4 suppresses osteoclast differentiation and osteoporosis by inhibiting autophagy through Ca2+ -calcineurin-NFATc1 pathway.
Knockdown of TRPV4 suppresses osteoclast differentiation and osteoporosis by inhibiting autophagy through Ca2+ -calcineurin-NFATc1 pathway.
Journal of cellular physiology (2018-11-06)
Boran Cao, Xin Dai, Wenbo Wang
RÉSUMÉ
The aim of this study is to evaluate the effect of transient receptor potential vanilloid 4 (TRPV4) on osteoclast differentiation and osteoporosis, and to investigate the underlying mechanism. The results showed that TRPV4 expression and intracellular Ca2+ concentration were significantly upregulated in macrophage colony-stimulating factor (M-CSF)-stimulated and receptor activator of nuclear factor κΒ ligand (RANKL)-stimulated RAW264.7 cells. Furthermore, TRPV4 overexpression further increased the M-CSF- and RANKL-induced number of tartrate-resistant acid phosphatase (TRAP)-positive osteoclasts and expression of osteoclastogenesis-related genes (TRAP, c-Fos, and nuclear factor of activated T cells [NFATc1]), activated the Ca 2+ -calcineurin-NFATc1 signaling and increased autophagy-related proteins (light chain [LC] 3II and Beclin-1) during osteoclast differentiation. In contrast, TRPV4 knockdown exerted the opposite effects. Mechanically, inhibition of Ca 2+ -calcineurin-NFATc1 signaling by FK506 or 11R-VIVIT abrogated the TRPV4 overexpression-induced osteoclast differentiation and autophagy induction. Moreover, suppression of autophagy by 3-methyladenine attenuated the TRPV4-induced osteoclast differentiation. In addition, short hairpin RNA TRPV4-lentivirus administration significantly diminished the increased levels of several osteoclastogenesis-related genes (RANKL, TRAP, and tumor necrosis factor-α), alleviated the disturbed microarchitecture of lumbar vertebrae, restored the decreased bone mineral density, ratio of bone volume to total tissue volume, trabecular thickness, and trabecular number, and diminished the increased trabecular separation, in ovariectomy (OVX)-induced osteoporosis mice. Consistent with the in vitro data, TRPV4 knockdown significantly decreased the induced number of TRAP-positive osteoclasts, the increased LC3 and NFATc1 expression in the lumbar vertebrae of OVX mice. In conclusion, TRPV4 knockdown suppresses osteoclast differentiation and osteoporosis by inhibiting autophagy through Ca 2+ -calcineurin-NFATc1 pathway.