MilliporeSigma
  • In Endothelial Cells, the Activation or Stimulation of Soluble Guanylyl Cyclase Induces the Nitric Oxide Production by a Mechanism Dependent of Nitric Oxide Synthase Activation.
In Endothelial Cells, the Activation or Stimulation of Soluble Guanylyl Cyclase Induces the Nitric Oxide Production by a Mechanism Dependent of Nitric Oxide Synthase Activation.
Journal of pharmacy & pharmaceutical sciences : a publication of the Canadian Society for Pharmaceutical Sciences, Societe canadienne des sciences pharmaceutiques (2018-02-17)
Ariane Migliato Martinelli, Carla Nascimento Dos Santos Rodrigues, Thiago Francisco de Moraes, Gerson Jhonatan Rodrigues
RÉSUMÉ
In endothelial cells, investigate if the soluble guanylate cyclase (sGC) activation or stimulation is able to potentiate the relaxation in vessels. Aortic and coronary rings with and without endothelium were placed in a myograph and cumulative concentration-effect curves for DETA-NO or ataciguat were performed. Nitric oxide (NO) were measured by fluorescence or by selective electrode in human umbilical endothelial cells (HUVECs) in response to some treatments, including ataciguat, 8-Br-cGMP and A23187. The presence of the endothelium potentiated the relaxation induced by DETA-NO in aortic and coronary rings. In addition, in aortic rings the endothelium potentiated the relaxation induced by ataciguat. In the presence of nitric oxide synthase (NOS) inhibitor, the endothelium effect was abolished to DETA-NO or ataciguat, in both vessels. Ataciguat, 8-Br-cGMP and A23187 were able to induce NO production in HUVECs cells. In the presence of NOS inhibitor, the NO production induced by ataciguat and 8-Br-cGMP was abolished. Our results suggest that in aortic and coronary rings the endothelium potentiates the relaxation induced by activation or stimulation of sGC through a mechanism dependent of NOS activation. This article is open to POST-PUBLICATION REVIEW. Registered readers (see "For Readers") may comment by clicking on ABSTRACT on the issue's contents page.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Ataciguat, ≥98% (HPLC)