MilliporeSigma
Photolytic labeling to probe molecular interactions in lyophilized powders.
Molecular pharmaceutics (2013-10-16)
Lavanya K Iyer, Balakrishnan S Moorthy, Elizabeth M Topp
RÉSUMÉ
Local side-chain interactions in lyophilized protein formulations were mapped using solid-state photolytic labeling-mass spectrometry (ssPL-MS). Photoactive amino acid analogues (PAAs) were used as probes and either added to the lyophilized matrix or incorporated within the amino acid sequence of a peptide. In the first approach, apomyoglobin was lyophilized with sucrose and varying concentrations of photoleucine (L-2-amino-4,4'-azipentanoic acid; pLeu). The lyophilized solid was irradiated at 365 nm to initiate photolabeling. The rate and extent of labeling were measured using electrospray ionization/high-performance liquid chromatography/mass spectrometry (ESI-HPLC-MS), with labeling reaching a plateau at ~30 min, forming up to six labeled populations. Bottom-up MS/MS analysis was able to provide peptide-level resolution of the location of pLeu. ssPL-MS was also able to detect differences in side-chain environment between sucrose and guanidine hydrochloride formulations. In the second approach, peptide GCG (1-8)* containing p-benzoyl-L-phenylalanine (pBpA) in the amino acid sequence was lyophilized with various excipients and irradiated. Peptide-peptide and peptide-excipient adducts were detected using MS. Top-down MS/MS on the peptide dimer provided amino acid-level resolution regarding interactions and the cross-linking partner for pBpA in the solid state. The results show that ssPL-MS can provide high-resolution information about protein interactions in the lyophilized environment.