MilliporeSigma
  • Production of n-octanoyl-modified ghrelin in cultured cells requires prohormone processing protease and ghrelin O-acyltransferase, as well as n-octanoic acid.
Production of n-octanoyl-modified ghrelin in cultured cells requires prohormone processing protease and ghrelin O-acyltransferase, as well as n-octanoic acid.
Journal of biochemistry (2009-07-25)
Tomoko Takahashi, Takanori Ida, Takahiro Sato, Yoshiki Nakashima, Yuki Nakamura, Akihiko Tsuji, Masayasu Kojima
RÉSUMÉ
Ghrelin was originally isolated from rat stomach as an endogenous ligand for the GH secretagogue receptor. The major active form of ghrelin is a 28-amino acid peptide modified by an n-octanoic acid on the serine 3 residue, and this lipid modification is essential for the biological activity of ghrelin. However, it is not clear whether prohormone convertase (PC) and ghrelin O-acyltransferase (GOAT) are the minimal requirements for synthesis of acyl-modified ghrelin in cultured cells. By using three cultured cell lines, TT, AtT20 and COS-7, in which the expression levels of processing proteases and GOAT vary, we examined the processing patterns of ghrelin precursor. We found that not only PC1/3 but also both PC2 and furin could process proghrelin to the 28-amino acid ghrelin. Moreover, the presence of PC and GOAT in the cells, as well as n-octanoic acid in the culture medium, was necessary to produce n-octanoyl ghrelin.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Octanoyl coenzyme A lithium salt hydrate, ≥95% (HPLC)