MilliporeSigma
5-aminolevulinic acid biosynthesis in Escherichia coli coexpressing NADP-dependent malic enzyme and 5-aminolevulinate synthase.
Journal of microbiology and biotechnology (2007-12-08)
Jeong-Ah Shin, Yeong Deok Kwon, Oh-Hee Kwon, Heung Shick Lee, Pil Kim
RÉSUMÉ
5-aminolevulinate (ALA) synthase (E.C. 2.3.1.37), which mediates the pyridoxal phosphate-dependent condensation of glycine and succinyl-CoA, encoded by the Rhodobacter sphaeroides hemA gene, enables Escherichia coli strains to produce ALA at a low level. To study the effect of the enhanced C4 metabolism of E. coli on ALA biosynthesis, NADP-dependent malic enzyme (maeB, E.C. 1.1.1.40) was coexpressed with ALA synthase in E. coli. The concentration of ALA was two times greater in cells coexpressing maeB and hemA than in cells expressing hemA alone under anaerobic conditions with medium containing glucose and glycine. Enhanced ALA synthase activity via coupled expression of hemA and maeB may lead to metabolic engineering of E. coli capable of large-scale ALA production.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Succinyl coenzyme A sodium salt, ≥85%