MilliporeSigma
Mice deficient for the CD40 ligand.
Immunity (1994-08-01)
J Xu, T M Foy, J D Laman, E A Elliott, J J Dunn, T J Waldschmidt, J Elsemore, R J Noelle, R A Flavell
RÉSUMÉ
To study the potential roles of CD40L in immune responses, we generated CD40L-deficient mice by gene targeting. Similar to the effects of CD40L mutations in humans (hyper-IgM syndrome), CD40L-deficient mice have a decreased IgM response to thymus-dependent antigens, fail altogether to produce an antigen-specific IgG1 response following immunization, yet respond normally to a T-independent antigen, TNP-Ficoll. Moreover, these mice do not develop germinal centers in response to thymus-dependent antigens, suggesting an inability to develop memory B cell responses. Although CD40L-deficient mice have low levels of most circulating immunoglobulin isotypes, they do not exhibit the spontaneous hyper-IgM syndrome seen in humans, at least up to 12 weeks of age. In summary, our study confirms the important role of CD40-CD40L interactions in thymus-dependent humoral immune responses and germinal center formation.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Kit d′analyse de la PTP n°1, PTP Assay Kit 1 can be used to detect PTP-1B activity by dephosphorylation of the phosphopeptide (RRLIEDAEpYAARG) using malachite green detection, or by hydrolysis of pNPP.