- Guilty as charged: B-RAF is a human oncogene.
Guilty as charged: B-RAF is a human oncogene.
Cancer cell (2004-10-19)
Mathew J Garnett, Richard Marais
PMID15488754
RÉSUMÉ
RAF proteins are components of a conserved signaling pathway that regulates cellular responses to extracellular signals. Despite over 20 years of research into the regulation and function of the RAF proteins, it was only realized recently that the B-RAF isoform is mutated at a high frequency in human cancer. Here we review the rapid progress made in our understanding of B-RAF as an oncogene and of its role in cancer.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
B-Raf (δ1-415) Protein, active, 10 µg, Active, N-terminal GST-tagged recombinant human B-Raf residues 416-end, for use in Kinase Assays.
Sigma-Aldrich
B-Raf (V599E) Protein, active, 10 µg, Active, N-terminal GST-tagged recombinant human B-Raf residues 416-end, containing a V599E mutation, for use in Kinase Assays.