Merck
Recruitment of the kainate receptor subunit glutamate receptor 6 by cadherin/catenin complexes.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2002-08-02)
Françoise Coussen, Elisabeth Normand, Cécile Marchal, Pierre Costet, Daniel Choquet, Mireille Lambert, René-Marc Mège, Christophe Mulle
RÉSUMÉ
Kainate receptors modulate synaptic transmission by acting either at presynaptic or at postsynaptic sites. The precise localization of kainate receptors as well as the mechanisms of targeting and stabilization of these receptors in neurons are largely unknown. We have generated transgenic mice expressing the kainate receptor subunit glutamate receptor 6 (GluR6) bearing an extracellular myc epitope (myc-GluR6), in forebrain neurons, in which it assembles with endogenous kainate receptor subunits. In transgenic mice crossed with GluR6-deficient mice, myc-GluR6 efficiently rescues the missing subunit. Immunoprecipitation of transgenic brain extracts with anti-myc antibodies demonstrates an interaction with cadherins, beta-catenin, and p120 catenin, as well as with the associated proteins calcium calmodulin-dependent serine kinase and Velis, but not with alpha-catenin. In glutathione S-transferase-pulldown experiments, beta-catenin interacts, although indirectly, with the last 14 aa of GluR6. Transfected myc-GluR6 colocalizes with beta-catenin at cell-cell junctions in non-neuronal cells. Finally, activation of N-cadherins by ligand-covered latex beads recruits GluR6 to cadherin/catenin complexes. These results suggest an important role for cadherin/catenin complexes in the stabilization of kainate receptors at the synaptic membrane during synapse formation and remodeling.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-KA2/GRIK5 (Kainate Receptor) Antibody, Upstate®, from rabbit
Référence
Conditionnement
Disponibilité
Prix
Quantité