MilliporeSigma
Regulation of AMPK by the ubiquitin proteasome system.
The American journal of pathology (2011-01-13)
Makhosazane Zungu, Jonathan C Schisler, M Faadiel Essop, Chris McCudden, Cam Patterson, Monte S Willis
RÉSUMÉ
The 5'-AMP-activated protein kinase (AMPK) functions as a metabolic fuel gauge that is activated in response to environmental stressors to restore cellular energy balance. In the heart, AMPK coordinates the activation of glucose and fatty acid metabolic pathways to ensure increased production of myocardial ATP when required, such as during cardiac ischemia/reperfusion and hypertrophy, causing an increase in AMPK activity that can be viewed as both protective and maladaptive. While we understand the basic regulation of AMPK activity by kinases, recent studies have introduced the concept that AMPK is regulated by other post-translational modifications, specifically ubiquitination. These studies reported that the ubiquitin ligase cell death-inducing DFFA-like effector a ubiquitinates the β subunit of AMPK to regulate its steady-state protein levels. Other investigators found that AMPK regulatory components, including the AMPK α subunit and AMPK kinases NUAK1 and MARK4, can be ubiquitinated with atypical ubiquitin chains. The USP9X-deubiquitinating enzyme was identified to remove ubiquitination from both NUAK1 and MARK4. Lastly, AMPK activation increases the expression of the ubiquitin ligases MAFBx/Atrogin-1 and MuRF1. These ubiquitin ligases regulate key cardiac transcription factors to control cardiomyocyte mass and remodeling, thus suggesting another mechanism by which AMPK may function in the heart. The relevance of AMPK ubiquitination in cardiac disease has yet to be tested directly, but it likely represents an important mechanism that occurs in common cardiac diseases that may be targeted for therapy.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Adenosine 5′-monophosphate sodium salt, from yeast, ≥99%
Sigma-Aldrich
Adenosine 5′-monophosphate disodium salt, ≥99.0% (HPLC)
Sigma-Aldrich
Adenosine 5′-monophosphate monohydrate, from yeast, ≥97%