MilliporeSigma
  • Differential effects of β-glucan on oxidative stress, inflammation and copper transport in two intestinal regions of large yellow croaker Larimichthys crocea under acute copper stress.
Differential effects of β-glucan on oxidative stress, inflammation and copper transport in two intestinal regions of large yellow croaker Larimichthys crocea under acute copper stress.
Ecotoxicology and environmental safety (2018-09-08)
Lin Zeng, Yong-Hong Wang, Chun-Xiang Ai, Jian-She Zhang
RÉSUMÉ
The aim of the present study was to evaluate investigate the effects of β-glucan on oxidative stress, inflammation and copper transport in two intestinal regions of large yellow croaker under acute copper stress. Fish were injected with β-glucan at a dose of 0 or 5 mg kg-1 body weight on 6, 4 and 2 days before exposed to 0 and 368 μg Cu L-1 for 48 h. Biochemical indicators (MDA, Cu content, MTs protein levels, Cu/Zn-SOD, CAT and iNOS activities), gene expressions of oxidative stresses (Cu/Zn-SOD, CAT, Nrf2, MTs and MTF-1), inflammatory responses (NF-κB, iNOS, IL-1β, IL-6 and TNF-α) and Cu transporters (ATP7A, ATP7B and CTR1) were determined. In the anterior intestine, β-glucan increased MTs levels, activities of Cu/Zn-SOD, CAT and iNOS, mRNA levels of MTs, CAT, iNOS, ATP7A and ATP7B, and reduced Cu content and CTR1 gene expression to inhibite Cu-induced MDA. But β-glucan had no effect on inflammatory gene expressions. In the mid intestine, β-glucan increased activities of Cu/Zn-SOD and iNOS, mRNA levels of Cu/Zn-SOD, CAT and iNOS to maintain MDA content. However, unlike the anterior intestine, β-glucan had no effect on Cu transporter gene expressions. Furthermore, transcription factors (Nrf2, NF-κB and MTF-1) paralleled with their target genes in the mid intestine, but no correlation was observed between NF-κB and IL-1β and TNF-α gene expressions in the anterior intestine. In conclusion, our results unambiguously showed that β-glucan induced oxidative stress, inflammation and copper transport were varied between the anterior and mid intestines of fish under Cu stress.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
β-D-Glucan from barley, ≥95% (HPLC)