MilliporeSigma
25-Hydroxycholesterols in innate and adaptive immunity.
Nature reviews. Immunology (2014-10-18)
Jason G Cyster, Eric V Dang, Andrea Reboldi, Tangsheng Yi
RÉSUMÉ
Cholesterol and components of the cholesterol biosynthetic pathway have fundamental roles in all mammalian cells. Hydroxylated forms of cholesterol are now emerging as important regulators of immune function. This involves effects on the cholesterol biosynthetic pathway and cell membrane properties, which can have antiviral and anti-inflammatory influences. In addition, a dihydroxylated form of cholesterol functions as an immune cell guidance cue by engaging the G protein-coupled receptor EBI2, and it is required for mounting adaptive immune responses. In this Review, we summarize the current understanding of the closely related oxysterols 25-hydroxycholesterol and 7α,25-dihydroxycholesterol, and the growing evidence that they have wide-ranging influences on innate and adaptive immunity.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Avanti
25-hydroxycholesterol, Avanti Research - A Croda Brand