MilliporeSigma
Genetic ablation of interacting with Spt6 (Iws1) causes early embryonic lethality.
PloS one (2018-09-13)
Arturo Orlacchio, Aaron E Stark, Claudia Foray, Foued Amari, Tyler Sheetz, Erika Reese, Anna Tessari, Krista La Perle, Dario Palmieri, Philip N Tsichlis, Vincenzo Coppola
RÉSUMÉ
IWS1 is an RNA-polymerase II (RNAPII)-associated transcription elongation factor whose biological functions are poorly characterized. To shed some light on the function of this protein at the organismal level, we performed a systematic tissue analysis of its expression and generated Iws1-deficient mice. A thorough immunohistochemical characterization shows that IWS1 protein is present in the nucleus of all cells in most of the examined tissues, with few notable exceptions. We also report that ablation of Iws1 consistently causes lethality at the pre-implantation stage with high expression of the gene in fertilized oocytes. In summary, we are providing evidence that Iws1 is expressed in all adult organs and it is an essential gene for mouse embryonic development.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Anti-IWS1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution