MilliporeSigma
IRF5 is increased in labouring myometrium and regulates pro-labour mediators.
Reproduction (Cambridge, England) (2018-07-15)
Ratana Lim, Gillian Barker, Martha Lappas
RÉSUMÉ
Preterm birth continues to be the leading cause of neonatal mortality and morbidities that can extend into adult life. Few treatment options stem from our incomplete understanding of the mechanisms of human labour and delivery. Activation of the inflammatory response in gestational tissues by inflammation and/or infection leads to the production of pro-inflammatory and pro-labour mediators, thus preterm birth. Interferon regulatory factor 5 (IRF5) has recently emerged as an important pro-inflammatory transcription factor involved in acute and chronic inflammation. The aims of this study were to determine the expression of IRF5 in human myometrium from labouring and non-labouring women, and whether IRF5 is involved in the genesis of pro-inflammatory and pro-labour mediators induced by pro-inflammatory cytokines or toll-like receptor (TLR) ligands. IRF5 mRNA and protein expression was significantly higher in human myometrium after spontaneous term labour, compared to non-labouring tissues. IRF5 mRNA expression was also significantly higher in primary myometrial cells treated with the pro-inflammatory cytokines IL1B or TNF. In primary myometrial cells, IRF5 knockdown by siRNA (siIRF5) was associated with significantly decreased expression and or secretion of pro-inflammatory cytokines (IL1A, IL6), chemokines (CXCL8, CCL2), adhesion molecules (ICAM1, VCAM1) and contraction-associated proteins PTGS2, PGF2α and PTGFR when in the presence of IL1B, TNF, fsl-1 (TLR2/6 ligand) or flagellin (TLR5 ligand). siIRF5-transfected cells also displayed decreased NF-κB RELA transcriptional activity in the presence of these preterm birth mediators. Our study suggests a novel role for IRF5 in the regulation of the inflammatory response in human myometrium.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
MISSION® esiRNA, targeting human IRF5