MilliporeSigma
  • Rearing Zebrafish on Black Soldier Fly (Hermetia illucens): Biometric, Histological, Spectroscopic, Biochemical, and Molecular Implications.
Rearing Zebrafish on Black Soldier Fly (Hermetia illucens): Biometric, Histological, Spectroscopic, Biochemical, and Molecular Implications.
Zebrafish (2018-03-29)
Arturo Vargas, Basilio Randazzo, Paola Riolo, Cristina Truzzi, Giorgia Gioacchini, Elisabetta Giorgini, Nino Loreto, Sara Ruschioni, Matteo Zarantoniello, Matteo Antonucci, Sara Polverini, Gloriana Cardinaletti, Simona Sabbatini, Francesca Tulli, Ike Olivotto
RÉSUMÉ
A desirable goal of the aquaculture sector is to replace most of fish meal and fish oil with more sustainable, cost-effective, and environmental friendly ingredients ensuring fish health and welfare standards. Due to minimal environmental impact, compared with most conventional feed commodities, insects deserve a growing attention as candidate ingredients for aquafeeds. The present study investigated, for the first time, the possible application of a 100% insect diet in zebrafish larval rearing. Through a multidisciplinary approach, the major biological responses of fish to the new diets were assessed. Results of biometry, fatty acid composition, expression of genes involved in fish growth, stress response, lipid metabolism, chitinolytic activity, gut inflammation, and liver macromolecular composition suggested a possible application of insect larvae for zebrafish larval rearing. However, further studies are necessary to better understand the use of this insect species in the rearing of fish.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
RNAzol® RT, For processing total and small RNA from human, animal, plant, bacterial,and viral samples