MilliporeSigma
The novel fungicide SYP-14288 acts as an uncoupler against Phytophthora capsici.
Pesticide biochemistry and physiology (2018-06-24)
Zhiwen Wang, Xiaoxia Ni, Qin Peng, Yanhua Hou, Yuan Fang, Wenjun Mu, Changling Liu, Pengfei Liu, Xili Liu
RÉSUMÉ
SYP-14288 is a novel fungicide developed by the Shenyang Research Institute of Chemical Industry in China. Although preliminary studies indicate that SYP-14288 is highly effective against 32 important plant pathogens belonging to a range of taxonomic groups, its mode of action remains unknown. In this study, we documented that SYP-14288 has excellent activity against all of the asexual life stages of the plant-pathogenic oomycete Phytophthora capsici, and is especially effective in blocking cyst germination and other life stages that require high energy consumption. In assays designed to determine the fungicide's mode of action, addition of ATP reduced SYP-14288 inhibition of P. capsici, which suggested that SYP-14288 inhibits ATP synthesis of the pathogen. This inference was confirmed in that treatment with SYP-14288 sharply reduced the ATP content in P. capsici. The respiration rate of P. capsici was positively correlated with the concentration of SYP-14288 or of the fungicide fluazinam (an uncoupler of oxidative phosphorylation), but increases in respiration were greater with SYP-14288 than with fluazinam. These results indicate that SYP-14288 is a promising fungicide that functions as an uncoupler of oxidative phosphorylation.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Supelco
Azoxystrobin, PESTANAL®, analytical standard