MilliporeSigma
Prostaglandin E2 as a Regulator of Immunity to Pathogens.
Pharmacology & therapeutics (2017-12-25)
Giovanny J Martínez-Colón, Bethany B Moore
RÉSUMÉ
The body is exposed to foreign pathogens every day, but remarkably, most pathogens are effectively cleared by the innate immune system without the need to invoke the adaptive immune response. Key cellular components of the innate immune system include macrophages and neutrophils and the recruitment and function of these cells are tightly regulated by chemokines and cytokines in the tissue space. Innate immune responses are also known to regulate development of adaptive immune responses often via the secretion of various cytokines. In addition to these protein regulators, numerous lipid mediators can also influence innate and adaptive immune functions. In this review, we cover one particular lipid regulator, prostaglandin E2 (PGE2) and describe its synthesis and signaling and what is known about the ability of this lipid to regulate immunity and host defense against viral, fungal and bacterial pathogens.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Sulprostone, ≥95% (HPLC), oil