MilliporeSigma
  • The olive oil phenolic (-)-oleocanthal modulates estrogen receptor expression in luminal breast cancer in vitro and in vivo and synergizes with tamoxifen treatment.
The olive oil phenolic (-)-oleocanthal modulates estrogen receptor expression in luminal breast cancer in vitro and in vivo and synergizes with tamoxifen treatment.
European journal of pharmacology (2017-06-20)
Nehad M Ayoub, Abu Bakar Siddique, Hassan Y Ebrahim, Mohamed M Mohyeldin, Khalid A El Sayed
RÉSUMÉ
Luminal breast cancer represents a therapeutic challenge in terms of aggressive disease and emerging resistance to targeted therapy. (-)-Oleocanthal has demonstrated anticancer activity in multiple human cancers. The goal of this study was to explore the effect of (-)-oleocanthal treatment on growth of luminal breast cancer cells and to examine the effect of combination of (-)-oleocanthal with tamoxifen. Results showed that (-)-oleocanthal inhibited growth of BT-474, MCF-7, and T-47D human breast cancer cells in mitogen-free media with IC50 values of 32.7, 24.07, and 80.93µM, respectively. Similarly, (-)-oleocanthal suppressed growth of BT-474, MCF-7, and T-47D cells in 17β-estradiol-supplemented media with IC50 values of 22.28, 20.77, and 83.91µM, respectively. Combined (-)-oleocanthal and tamoxifen treatments resulted in a synergistic growth inhibition of BT-474, MCF-7, and T-47D cells with combination index values of 0.65, 0.61, and 0.53 for each cell line, respectively. In-silico docking studies indicated high degree of overlapping for the binding of (-)-oleocanthal and 17β-estradiol to estrogen receptors, while (-)-oleocanthal and tamoxifen have distinguished binding modes. Treatment with 5mg/kg or 10mg/kg (-)-oleocanthal resulted in 97% inhibition of tumor growth in orthotopic athymic mice bearing BT-474 tumor xenografts compared to vehicle-treated animals. (-)-Oleocanthal treatment reduced total levels of estrogen receptors in BT-474 cells both in vitro and in vivo. Collectively, (-)-oleocanthal showed a potential beneficial effect in suppressing growth of hormone-dependent breast cancer and improving sensitivity to tamoxifen treatment. These findings provide rational for evaluating the effect of (-)-oleocanthal in combination with endocrine treatments in luminal breast cancer.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Oleocanthal, from Olea europaea