MilliporeSigma
Molecular cloning and characterization of prokineticin receptors.
Biochimica et biophysica acta (2002-11-13)
Takatoshi Soga, Shun ichiro Matsumoto, Tamaki Oda, Tetsu Saito, Hideki Hiyama, Jun Takasaki, Masazumi Kamohara, Takahide Ohishi, Hitoshi Matsushime, Kiyoshi Furuichi
RÉSUMÉ
Recent studies have identified two novel biofunctional proteins, termed prokineticin 1/EG-VEGF and prokineticin 2, which were mammalian homologues of mamba MIT1 and frog Bv8. Prokineticins have been demonstrated to exert their physiological functions through G-protein coupled receptors (GPCRs). In this study, we report the molecular identification of two endogenous prokineticin receptors, designated PK-R1 and PK-R2, through a search of the human genomic DNA database. PK-R1, locating in chromosome 2, and PK-R2, locating in chromosome 20p13, shared 87% homology, which was an extremely high value among known GPCRs. In functional assays, mammalian cells expressing PK-Rs responded to prokineticins in a concentration-dependent manner. Tissue distribution analysis revealed that expression of PK-R1 was observed in the testis, medulla oblongata, skeletal muscle and skin, while that of PK-R2 showed preferential expression in the central nervous system. The tissue distribution of PK-Rs reported in this paper suggests that the prokineticins play multifunctional roles in vivo.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Prokineticin-2 human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture