MilliporeSigma
Effects of fatty acid glyceride product SILOhealth 104 on the growth performance and carcass composition of broiler chickens.
Poultry science (2018-02-21)
A Bedford, H Yu, M Hernandez, E J Squires, S Leeson, J Gong
RÉSUMÉ
Butyric acid is the primary energy source for colonocytes, and has shown potential as an alternative to in-feed antibiotics, due to its antimicrobial activity and positive effects on production performance traits of broiler chickens. SILOhealth 104 (SILO S.P.A., Florence, Italy) is a commercial product mainly containing mono- and di-glycerides of butyrate with a small portion of propionic, caprylic, capric, and lauric acid mono- and di-glycerides. Its effects on broiler performance and carcass composition have yet to be evaluated. Four-hundred-eighty day-old male Ross 308 birds were divided into different dietary treatment groups with equal starting weights and fed a diet containing 0, 500, 1,000, 2,000, or 3,000 ppm of SILOhealth 104 for 35 days. There were no significant differences in overall average daily gain or feed: gain ratio with the addition of SILOhealth 104 to the diets (P > 0.05). At 5 wk of age, abdominal fat weight was reduced in birds supplemented with SILOhealth 104 in a dose-responsive manner (P < 0.05), while breast muscle weight increased with supplementation, with significant increases in 2,000 ppm and 3,000 ppm birds compared to controls (P < 0.05). A significant reduction in gene expression of both forkhead box protein O4 and myostatin, 2 factors that can inhibit protein synthesis, was found in the breast muscle of all SILOhealth 104 treated birds (P < 0.05). In addition, gene expression in the adipose tissue, including acetyl-CoA carboxylase alpha and lipoprotein lipase, which are associated with lipid metabolism, was significantly decreased and increased, respectively, by the supplementation of SILOhealth 104 (P < 0.05). These data suggest that the components of SILOhealth 104 can positively affect the deposition of muscle, while reducing abdominal fat deposition in broiler chickens.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
1-monobutyrin, AldrichCPR