MilliporeSigma
Hepatitis B virus core protein promotes hepatocarcinogenesis by enhancing Src expression and activating the Src/PI3K/Akt pathway.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2018-02-07)
Wei Liu, Teng-Fei Guo, Zhen-Tang Jing, Zhi Yang, Lei Liu, Yuan-Ping Yang, Xu Lin, Qiao-Yun Tong
RÉSUMÉ
Hepatitis B virus core protein (HBc) is expressed preferentially in hepatitis B virus (HBV)-associated hepatocellular carcinoma (HCC). HBc can function as an oncogene arising from its gene regulatory properties, but how it contributes functionally to hepatocarcinogenesis remains unclear. In this study, we determined the molecular and functional roles of HBc during HBV-associated hepatocellular tumorigenesis. HBc increased tumor formation of hepatoma cells. Moreover, expression of HBc specifically promoted proliferation of hepatoma cells in vitro. Mechanistic investigations revealed that these effects were caused by activation of the Src/PI3K/Akt pathway through proximal switch from inactive Src to the active form of the kinase by HBc. HBc-mediated sarcoma (Src) kinase activation was associated with down-regulation of C-terminal Src kinase (Csk). In addition, HBc enhances Src expression by activation of alternative Src 1A promoter in an Sp1 transcription factor-dependent manner. Proliferation induced by stable HBc expression was associated with increased G1-S cell cycle progression mediated by Src kinase activation. HBc-induced cellular proliferation and tumor formation were reversed by administration of the Src inhibitor saracatinib. Together, our findings suggest that HBc promotes tumorigenesis of hepatoma cells by enhancing the expression of total Src and the active form of the kinase and subsequently activates Src/PI3K/Akt signaling pathway, revealing novel insights into the underlying mechanisms of HBV-associated hepatocarcinogenesis.-Liu, W., Guo, T.-F., Jing, Z.-T., Yang, Z., Liu, L., Yang, Y.-P., Lin, X., Tong, Q.-Y. Hepatitis B virus core protein promotes hepatocarcinogenesis by enhancing Src expression and activating the Src/PI3K/Akt pathway.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Src