MilliporeSigma
A Nonaqueous Potassium-Based Battery-Supercapacitor Hybrid Device.
Advanced materials (Deerfield Beach, Fla.) (2018-04-01)
Ling Fan, Kairui Lin, Jue Wang, Ruifang Ma, Bingan Lu
RÉSUMÉ
A low cost nonaqueous potassium-based battery-supercapacitor hybrid device (BSH) is successfully established for the first time with soft carbon as the anode, commercialized activated carbon as the cathode, and potassium bis(fluoro-slufonyl)imide in dimethyl ether as the electrolyte. This BSH reconciles the advantages of potassium ion batteries and supercapacitors, achieving a high energy density of 120 W h kg-1 , a high power density of 599 W kg-1 , a long cycle life of 1500 cycles, and an ultrafast charge/slow discharge performance (energy density and power density are calculated based on the total mass of active materials in the anode and cathode). This work demonstrates a great potential of applying the nonaqueous BSH for low cost electric energy storage systems.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
PTCDA, 97%