MilliporeSigma
Cr/Ni-catalyzed vinylation of aldehydes: a mechanistic study on the catalytic roles of nickel and chromium.
Chemistry (Weinheim an der Bergstrasse, Germany) (2011-03-23)
Wacharee Harnying, André Kaiser, Axel Klein, Albrecht Berkessel
RÉSUMÉ
The roles of nickel and chromium catalysts in the coupling reaction of vinyl halides and aldehydes, the so-called Nozaki-Hiyama-Kishi (NHK) reaction, have been studied by UV/Vis spectroscopy, electrochemical, and spectroelectrochemical methods. Electrochemical studies revealed that nickel plays the central role in activating the vinyl halide by reductive cleavage, to form a rapidly decomposing vinyl-Ni species. The latter can, however, be stabilized in the presence of the Cr complex. The redox behavior of the Ni complexes in the presence of vinyl halide demonstrated that the vinyl halide activation results from interaction with a one-electron reduced nickel species [formally Ni(I) ], not necessarily with a Ni(0) species. It was furthermore shown by UV/Vis spectroscopy and spectroelectrochemical methods that low-valent nickel [Ni(0) ] results from the interaction of the Ni(II) catalyst with CrCl(2) .
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
[4,4′-Bis(1,1-dimethylethyl)-2,2′-bipyridine] nickel (II) dichloride