MilliporeSigma
  • Effects of catechol-O-methyltransferase inhibitors and L-3,4-dihydroxyphenylalanine with or without carbidopa on extracellular dopamine in rat striatum.
Effects of catechol-O-methyltransferase inhibitors and L-3,4-dihydroxyphenylalanine with or without carbidopa on extracellular dopamine in rat striatum.
Journal of neurochemistry (1993-01-01)
S Kaakkola, R J Wurtman
RÉSUMÉ
The effects of two new catechol-O-methyltransferase (COMT) inhibitors, OR-611 and Ro 40-7592, in combination with L-3,4-dihydroxyphenylalanine (L-dopa) with or without carbidopa on extracellular levels of dopamine (DA), 3,4-dihydroxyphenylacetic acid (DOPAC), homovanillic acid (HVA), 3-O-methyldopa (3-OMD), and 5-hydroxyindoleacetic acid in rat striatum were studied. A dose of 10 mg/kg i.p. of Ro 40-7592 alone, in contrast to the same dose of OR-611, decreased the dialysate level of HVA and increased that of DOPAC; this dose was thus used to differentiate between the effects of central and peripheral COMT inhibition. L-Dopa (50 mg/kg i.p.) alone slightly increased extracellular levels of DA, DOPAC, and HVA. The effects of L-dopa were potentiated by carbidopa (50 mg/kg i.p.), and even 3-OMD levels in dialysate samples became detectable. Both OR-611 and Ro 40-7592 significantly further increased the DA and DOPAC efflux from striatum produced by L-dopa. This increase was more pronounced when carbidopa was added to the treatment. OR-611 did not modify the effect of L-dopa or carbidopa/L-dopa on dialysate HVA levels, whereas Ro 40-7592 markedly reduced those levels. Both OR-611 and Ro 40-7592 very clearly suppressed dialysate 3-OMD levels produced by carbidopa/L-dopa. Ro 40-7592 was more effective than OR-611 in potentiating the effects of L-dopa or carbidopa/L-dopa. These in vivo data show that the new COMT inhibitors markedly inhibit the O-methylation of L-dopa and increase its availability to brain, which is reflected as increased DA formation.(ABSTRACT TRUNCATED AT 250 WORDS)
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
S-(−)-Carbidopa, ≥98% (TLC), powder