Documents
Z659304
Bandelin SONOPULS ultrasonic homogenizer accessories
Rosett cell RZ 3
Synonyme(s) :
Glass sample vessel
About This Item
Code UNSPSC :
41101520
Nomenclature NACRES :
NB.74
Matériaux
glass
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bandelin 522
Disponibilité
available only in EU
Volume × D.I. × H
120 mL × 50 mm × 135 mm
Description générale
A rosette cell (RZ) enables intensive and uniform sonication of liquid media. The round shape of the side arms leads to an intensified circulation of the sample.
Caractéristiques et avantages
  • Optimal fluid circulation .
  • Good heat dissipation .
  • Homogeneous sound reinforcement results