Key Documents
MTCH6800100
MTC Bio Halo Lightweight Single Channel Pipettor
capacity 10-100 μL
Synonyme(s) :
pipette
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.15
Conditionnement
pack of 1 ea
Fabricant/nom de marque
MTC Bio H6800-100
Capacité
10-100 μL
Description générale
Halo Single Channel Pipettor, lightweight, 0.5-10µL
Caractéristiques et avantages
  • 10-40% lighter than leading competitors
  • 4 Digit micrometer with DriftGuard protection
  • Exclusive 1250µL model for extended length tips
  • User-enabled calibration port
  • Compatible with universal tips
Informations légales
Halo is a trademark of MTC Bio
MTC is a trademark of MTC Bio