Documents
Z569658
KDS syringe multi-rack for Legato series pumps
small
Synonyme(s) :
dosing pump syringe rack, infusion pump syringe rack
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.01
Conditionnement
pack of 1 ea
Fabricant/nom de marque
KD Scientific 78-8300
Description générale
Accommodates up to 6 x 30 to 60 mL syringes or up to ten 0.5 μL to 20 mL. Works with Legato 200, 210 and 210P.
Informations légales
Legato is a trademark of KD Scientific