Principaux documents
Z288284
Sigma-Aldrich
Mimi-pompe KNF Laboport®
Pump only, AC/DC input 115 V AC
Synonyme(s) :
laboratory vacuum pump, mini diaphragm vacuum pump, mini vacuum pump, vacuum pump
About This Item
Code UNSPSC :
41105101
Nomenclature NACRES :
NB.79
Caractéristiques
CE compliant
CSA approved
Conditionnement
pack of 1 ea
Fabricant/nom de marque
KNF
Disponibilité
not available in EU
Paramètres
35 psig max. pressure
5.5 L/min pumping speed
~120 torr vacuum
Poids
4.2 lb
Description générale
The ideal pump for vacuum and pressure filtration, solid phase extraction, and blotting. High performance diaphragm vacuum/pressure pumps for moderately corrosive applications and any filtration or procedure that requires clean evacuation, transfer and compression of air, gases, and vapors. Pumps are available with gauges and regulators for precise control of vacuum and pressure.
  • Portable
  • Oil-free
  • Quiet operation
  • Maintenance-free, Ryton® pump head
  • Molded PTFE diaphragm
  • Kalrez® multi-port valves
  • Single stage
Informations légales
Kalrez is a registered trademark of E. I. du Pont de Nemours and Company
Laboport is a registered trademark of KNF Neuberger GmbH
Ryton is a registered trademark of Chevron Phillips Chemical Company LP