Key Documents
THSD3144
ThalesNano MidiCart catalyst cartridge system
L × diam. 116 mm × 9.2 mm, nickel sponge, 1% Mo
Synonyme(s) :
Heterogeneous catalysis system, safe Heterogeneous catalysis
About This Item
Code UNSPSC :
41103817
Nomenclature NACRES :
NB.51
Conditionnement
pack of 1 ea
Fabricant/nom de marque
ThalesNano THS D3144
L × diam.
116 mm × 9.2 mm
Description générale
MiDiCarts are suitable for use only with Thalesnano H-Cube MidiCart reactors.
Informations légales
MidiCart is a trademark of ThalesNano Nanotechnology Inc.