Documents
SYNS310217
Sigma-Aldrich
Synthware T-bore glass stopcock Entire assembly
bore size 2 mm, plug size 12/30
Synonyme(s) :
Synthware S310217, t-shape stopcock
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware S310217
Taille de l'alésage
2 mm
Description générale
With solid glass plug. With constant pressure retention spring and clip.
Informations légales
Synthware is a trademark of Kemtech America Inc.