Documents
SYNF214000
Synthware four-neck round-bottom flask with vertical side necks
capacity 1,000 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40
Synonyme(s) :
Synthware F214000, 4-neck reaction flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
Matériaux
borosilicate glass flask
four-neck
round bottom
Joint
center joint: ST/NS24/40
side joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F214000
Capacité
1,000 mL
Description générale
With identical vertical side joints.
Informations légales
Synthware is a trademark of Kemtech America Inc.