Documents
CLS618075
Pyrex® fluted funnel, short stem
stem L 75 mm
Synonyme(s) :
Corning® 6180
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.43
Fabricant/nom de marque
Corning 6180-75
L de la tige
75 mm
D.E. de la tige
8-9 mm
D.I. de la partie supérieure
75 mm
Description générale
Beaded edge. Has depressed inside flutings to decrease filtering time. 60° angle cone.
Informations légales
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.