Troubleshooting Guide for Duolink® PLA for Flow Cytometry Detection
The combination of Duolink® PLA technology and flow cytometry allows quantitative detection of protein-protein interactions, post-translational modifications, and protein expression with greater statistical power. In order to get the best results, a lot of planning, prep work, and troubleshooting is required. This guide provides general tips and tricks for proper experiment execution, aids in identifying potential problems, and provides solutions to ensure a successful Duolink® PLA experiment for flow cytometry.
General Tips & Tricks
Troubleshooting
Frequently Asked Questions
Tip & Tricks
This section highlights general considerations and delineates important reminders to help your Duolink® PLA experiment for flow cytometry proceed smoothly.
Upfront Considerations
- Use conditions suitable for optimal primary antibody performance within the sample to be tested. These include sample processing parameters (fixation and permeabilization), primary antibody titer, and primary antibody incubation conditions (temperature and duration). When two primary antibodies are used for a Duolink® PLA experiment, make sure the sample processing conditions are compatible for optimal performance of both primary antibodies.
- Titration of primary antibody should be determined first by immunofluorescence (IF) by traditional flow cytometry (preferable) or microscopy. It is recommended to use indirect detection during initial optimization as directly conjugated antibodies may not represent the appropriate amount of antibody needed. When done properly, minimal additional optimization is required when applied to Duolink® PLA technology.
- Duolink® Blocking Solution and Antibody Diluent have been optimized for use with Duolink® PLA reagents to minimize non-specific binding of the PLA probes and detection oligos. If alternative solutions have been optimized for primary antibody performance by traditional flow cytometry, it is likely these can be used in a Duolink® PLA experiment but will need to be determined by the user.
- If combining Duolink® PLA technology with IF for multicolor flow cytometry, choose fluorophores that minimize spectral overlap and reserve the brightest fluorophore option for the lowest expressing signal.
- Always include both technical and, when possible, biological controls in your experiment to properly evaluate the results.
Common Experimental Parameters
- Make sure to remove excess wash solutions from samples. Residual wash buffer can cause further dilution of antibodies and/or decrease ligation or amplification efficiency.
- Perform all steps at the appropriate temperatures and incubation times for best results, in particular the enzymatic steps (ligation and amplification).
- For detection of low-abundant proteins, extended amplification and/or detection times may be required.
- Perform washes in ample wash buffer at room temperature.
- Make sure ligation, amplification, and detection buffers are completely thawed and vortexed prior to usage.
Storage Conditions
- All Duolink® PLA probes must be stored at 2 - 8°C; this includes those generated using Duolink® Probemaker (PLUS or MINUS). Do not freeze PLA probes or results will significantly decrease.
- Store all Duolink® flowPLA Detection Reagents at -20°C. Keep enzymes in a freezer block while in use.
- Do not store diluted Duolink® PLA reagents.
- Resuspend cells in 1x PBS following the final wash step prior to analysis. Cells can be analyzed immediately or stored in the dark at 2 - 8°C for up to a week.
Figure 1.The Duolink PLA signal is stable. The same cells stored for a week give the same result as those immediately analyzed.
Data Analysis
- Duolink® flowPLA detection reagents should be selected based on the lasers and detectors in your instrument. Duolink® flowPLA fluorophore specifications are shown in Table 1.
- If combining Duolink® PLA technology with IF for multicolor flow cytometry, choose fluorophores that minimize spectral overlap and reserve the brightest fluorophore option for the lowest expressing signal.
- Use forward and side scatter parameters to gate on fixed cells and to remove cellular debris.
- Be sure to collect enough events during analysis, especially in cases of rare events, to get significance and an accurate population spread.
- Properly gate on controls to ensure correct positive signal.
Product No. | Reagent | Excitation (nm) | Excitation Laser Line (nm)* | Emission (nm) |
---|---|---|---|---|
DUO94005 | Duolink flowPLA Detection Violet | 390 | 405 | 476 |
DUO94002 | Duolink flowPLA Detection Green | 495 | 488 | 527 |
DUO94003 | Duolink flowPLA Detection Orange | 554 | 532, 561 | 579 |
DUO94001 | Duolink flowPLA Detection Red | 594 | 532, 561 | 624 |
DUO94004 | Duolink flowPLA Detection Far-Red | 644 | 638, 640, 642 | 669 |
*The excitation laser line represents commonly used lasers that excite the fluorophore. It does not necessarily reflect the lasers available for each particular instrument.
Troubleshooting
Despite all precautions, sometimes the results are not as expected. Lack of signal in a positive control or high background are two of the most common issues when performing immunodetection experiments, and may also occur with the Duolink® PLA technology. The tables below list the probable causes and suggested solutions to get around these common problems.
Too High of Background Signal
Probable Causes | Suggested Solutions |
---|---|
Concentration of primary antibody is too high |
|
Non-specific binding of primary antibodies |
|
Insufficient blocking |
Figure 2. Addition of salmon sperm DNA to the Duolink® flowPLA Detection Buffer can decrease background. |
Insufficient washing |
|
Drying of sample |
|
Precipitate in ligation buffer |
|
Dust, salt, or fixation precipitates cause highly fluorescent particles |
|
Probable Causes | Suggested Solutions |
---|---|
Inappropriate reaction volume |
![]() Figure 3. Varying the cell number per reaction can affect results. |
No or insufficient binding of primary antibodies |
|
Inappropriate dilutions of Duolink® PLA reagents |
Figure 4. Further dilution of Duolink® flowPLA reagents will adversely affect results. |
Inappropriate storage of Duolink® PLA reagents |
|
Inappropriate incubation temperatures |
|
Inefficient ligation |
|
Inefficient amplification or detection |
Figure 5. Increased amplification time can aid in the detection of low-abundant targets. |
Wash buffers used incorrectly |
|
Wrong filter used for acquisition |
|
Probable Causes | Suggested Solutions |
---|---|
Cell Loss |
|
Cell Clumping |
|
Probable Causes | Suggested Solutions |
---|---|
Autofluorescence |
|
Insufficient cell counts |
|
PMT voltages set incorrectly |
|
Coalescence of Signal |
Figure 6. Extended amplification times can lead to coalescence of PLA signals. |
Frequently Asked Questions
Refer to the Duolink® PLA Troubleshooting Guide for a list of frequently asked questions around the Duolink® PLA technology.
Network error: Failed to fetch