MilliporeSigma
HomeOrganic ElectronicsOrganic Semiconductor Laser Materials
Organic Semiconductor Laser Materials
Chihaya Adachi, Hajime Nakanotani
Kyushu University
Abstract
A wide variety of challenges aiming for electrical pumping of organic laser diodes have been addressed. However, organic laser diodes have difficulty gaining widespread application due to their high lasing thresholds under electrical pumping. A variety of organic semiconductors have been developed to reduce the lasing threshold, which include not only small molecular materials but also conjugated polymers. In this article, we demonstrate that bis-styrylbenzene derivatives show promising characteristics for low-lasing thresholds and discuss the design considerations for organic lasing molecules.
Introduction
The dissolution of luminescent organic dye molecules in an organic solvent followed by their excitation with high-energy light (greater than their lasing threshold) produces a laser beam. Due to their broad tunabilities, dye lasers have made remarkable advancements.1-5 In recent years, organic solid-state lasers involving the dispersion of an organic dye within a solid/crystal matrix have also been actively studied. In 1989, we proposed an idea to create an organic semiconductor laser diode using triplet excitons of rare earth complexes such as a europium (Eu) complex.6 This system would produce an electrically excited laser with a low threshold by forming a population inversion using the long excitation lifetime of Eu. There have been many studies reporting amplified spontaneous emission (ASE) oscillation from an organic solid thin film and laser oscillation from an organic thin film with an oscillator structure. The thin films in these studies were made by doping a polymer or small-molecule medium with an appropriate laser dye.7-11 These studies provide evidence that organic dyes have suitably high stimulated emission coefficients and that the formation of a solid thin film waveguide can provide the necessary architecture for a solidstate device with a low lasing threshold. More recently, laser oscillations from organic semiconductor dyes, which take electrical excitation into consideration, have also been actively studied.12,13 This interest was generated because organic light-emitting diodes (OLEDs) have achieved an internal luminescence efficiency of nearly 100% through the use of a triplet excited state at the luminescence center. This excited state leads to the realization of an organic semiconductor laser that is an extension of OLEDs a practical study subject of organic electronics.14 In addition, light-emitting transistors have also become capable of producing highly efficient electroluminescence (EL), prompting new electrical excitation developments of organic semiconductors.14-21 In this article, we will discuss materials issues for the production of an organic semiconductor laser. This process requires the development of a new laser material with a focus on a low lasing threshold. Derivatives of bis-styrylbenzene (BSB) have demonstrated extremely good ASE oscillation characteristics in organic solid thin films.15,16 As a result, we will introduce new organic semiconductor materials with similar ASE characteristics in organic solid thin films by using BSB analogs as active materials.
The Mechanism and Goal of Organic LEDs
The external quantum efficiency (ηext) of OLEDs is based on four factors: (i) the ratio of electron and hole injection, transport, and recombination (ν), (ii) exciton formation efficiency (ηr), (iii) internal electroluminescence quantum yield (φp), and (iv) light out-coupling efficiency (ηp) (Figure 1).
OLED luminescence processes (carrier injection, transport, recombination, exciton formation/decay, and light extraction) and their efficiencies. Figures in parentheses represent refractive indices.
Figure 1.OLED luminescence processes (carrier injection, transport, recombination, exciton formation/decay, and light extraction) and their efficiencies. Figures in parentheses represent refractive indices.
Obtaining the highest possible luminescence efficiency requires bringing each of the four factors as close to 100% as possible. Here, ν is a physical quantity containing the ratio of electron and hole injection and transport, as well as the probability of electron and hole recombination. φp can have a value near to 100% by using a material with a high internal luminescence quantum yield. Although ηr is generally low (25% when fluorescent materials are used), the use of phosphorescent materials can theoretically provide an ηr of 75 to 100%. If a device is formed on a common glass substrate, ηp is low at about 20%. Therefore, if fluorescent materials are used as luminescent molecules, ηext is approx 5% at most, but with the use of a triplet exciton as a luminescence transition process can provide a luminescence efficiency theoretically 3 or more times higher. In instances where the intersystem crossing (ISC) probability is about 100%, a luminescence efficiency approximately 4 times higher can be acheived. Figure 2 shows typical luminescent materials used in OLEDs.
Fluorescent and phosphorescent materials used in OLEDs
Figure 2.Fluorescent and phosphorescent materials used in OLEDs
For Alq3 (Product No. 444561) and DPVBi, luminescence is found during the transition from the excited singlet state to the ground state. In contrast, for Eu (Product No. 538957), platinum (Pt) (Product No. 673625), and iridium (Ir) (Product No. 688096) complexes, all singlet excitons undergo intersystem crossing into triplet excitons due to the internal heavy atom effect. In the triplet state, a radiative transition occurs with high luminescence efficiency. Use of an Ir compound at the luminescence center reportedly provides a high external EL quantum yield (ηext) of greater than 20%, showing that the internal quantum yield (ηint) reaches almost 100%.22 As described above, the electrons and holes injected into the organic layer result in almost immediate exciton formation.
Laser Active Materials (Fluorescent and Phosphorescent Materials)—Population Inversion Formation at a Low Threshold
To investigate various organic laser materials, we synthesized a variety of styrylamine-, coumarin-, and cyanine-based materials. Here, we will discuss the light amplification characteristics (ASE characteristics) of styrylbenzene derivatives, which we believe to be the most promising laser materials.15,16 Figure 3 presents a selection of the molecules that were studied.
Styrylbenzene-based organic laser materials
Figure 3.Styrylbenzene-based organic laser materials
Table 1 shows the ASE characteristics of thin films that are 6 wt% doped with styrylbenzene-based luminescent materials under optical excitation with 4,4’-bis[9-dicarbazolyl]-2,2’-biphenyl (CBP) (Product No. 660124) as the host. Typical BSB which have a dimer skeleton often exhibit especially low threshold (Eth ≅ 1.0 μJ/cm2). Molecules with a heterocycle have an ASE threshold of 20 to 100 μJ/cm2, and molecules with an azomethine skeleton do not cause emission spectrum narrowing even with an excitation energy >100 μJ/cm2. To identify the controlling factors of ASE thresholds, data values for absolute photoluminescence quantum efficiency (φf ), fluorescence lifetime (τf ), radiative decay constant (kr), and triplet-triplet absorption are shown in Table 1.
Table 1.Basic optical physical characteristics of styryl-based laser materials
The ASE thresholds are not directly correlated with φf or τf but are greatly correlated with kr. The magnitude of kr can determine how large the threshold is. 4,4’-bis[(N-carbazole)styryl]biphenyl (BSB-Cz) has extremely good ASE characteristics. The threshold of BSB-Cz is the lowest of the styryl-based fluorescent materials studied so far.15 The luminescence intensity and lifetime do not depend on temperature, so the nonradiative decay is completely inhibited, even at room temperature. The ASE oscillation wavelength of BSB-Cz corresponds to the 0-1 transition in the PL spectrum. The radiative decay constant (kr), stimulated emission cross-section (σem), and absorption crosssection (σABS) are calculated as below. kr (= ηPL/τf ) is calculated with the fluorescence lifetime (τf ) and luminescence quantum efficiency (ηPL) of each codeposited thin film. σem is given by equations (1) and (2) below with the absolute luminescence quantum yield and fluorescence lifetime of each codeposited thin film:24,25
where, Ef (λ) is quantum yield distribution, n (λ) is the refractive index of each wavelength, and τf is fluorescence lifetime. When the refractive index n = 1.8, the absorption cross-section σABS 26 is given by equation (3) below:
where, ε(λ) is the molar absorption coefficient, and NA is Avogadro’s number. kr and σem are strongly correlated with the ASE oscillation threshold Eth in each codeposited thin film. The ASE oscillation thresholds in thin films with the dimmers BSB-Cz, BSB-Me, and BSBOMe as active materials are 1 μJ/cm2 or less. The 6 wt%-BSB-Cz:CBP thin film’s σem is the highest. Although the SBD-4:CBP codeposited thin film is higher in kr and σem than the SBD-1:CBP codeposited thin film, SBD-4:CBP has a high oscillation threshold Eth and the saturation of luminescence intensity is noted at high excitation intensity. This suggests that there is a factor inhibiting luminescence in the 6 wt%-SBD-4:CBP codeposited thin film. Here, the effective stimulated emission cross-section is the difference between the stimulated emission cross-section and loss-related cross-sections (absorption cross-section and excited-state absorption cross-sections) and is given by equation (4) below:
where, σemeff is the effective stimulated emission cross-section, σem is the stimulated emission cross-section, σSS is the singlet excited-state absorption cross-section, and σTT is the triplet excited-state absorption cross-section. The absorption cross-section at the ASE oscillation wavelength of the SBD-4:CBP codeposited thin film is negligible, meaning that the excited-state absorption has a great impact on σemeff. Singlet and triplet excited-state absorption is found in any of the codeposited thin films, and the ASE oscillation wavelength of SBD-1 has triplet excited-state absorption, but not singlet excited-state absorption. In contrast, SBD-4 has singlet and triplet excited-state absorption in the luminescence wavelength range. This result shows that the occurrence of singlet excitation absorption under strong excitation inhibits the formation of population inversion, resulting in no stimulated emission. Figure 4 shows the stimulated emission and absorption cross-sections spectra and excited-state absorption spectrum of the 6 wt%-BSB-Cz:CBP codeposited thin film that has the lowest ASE oscillation threshold. For BSB-Cz, neither the single nor triplet excited-state absorption exists at the ASE oscillation wavelength, providing materials designed to perform at low thresholds.
ASE spectrum and S-S and T-T absorption spectra of BSB-Cz
Figure 4.ASE spectrum and S-S and T-T absorption spectra of BSB-Cz
Development to An Electrically Excitable Device Structure
OLED and FET structures also show promise in realizing an organic semiconductor laser. The OLED structure is basically a double heterostructure to effectively trap electrons and holes as well as trapping generated excitons within the luminescent layer. Advantages of this structure include the ability to use the OLED design guidelines developed so far and the flexibility to select a wide range of organic materials. However, a drawback of this structure is the failure to guide light effectively if a metal electrode is used as the cathode. Therefore, a transparent electrode such as ITO needs to be used at both the anode and cathode. For example, when a transparent device structure with ITO (30 nm)/a-NPD (20 nm)/CBP:BSB [6 wt%] (70 nm)/BCP (20 nm)/Alq3 (20 nm)/MgAg (X nm)/ITO (30 nm)15 is used, a clear ASE oscillation is noted under optical excitation as long as the MgAg layer is between 1 and 3 nm thick (Figure 5).
ASE characteristics under the excitation of light from an electrically excitable laser structure
Figure 5.ASE characteristics under the excitation of light from an electrically excitable laser structure
Recently, even transistor structures have allowed luminescent devices to be constructed. Light emission has actually been observed near the drain electrode by using p-type organic semiconductors such as oligothiophene27, tetracene28 (Product No. 698415), fluorene-based polymers (Product No. 571032), PPV-based polymers29 (Product No. 541443), and thiophene-based oligomers30 (Figure 6).
Organic semiconductor materials for light-emitting transistors
Figure 6.Organic semiconductor materials for light-emitting transistors
Also, use of a tetraphenylpyrene derivative (TPPy) has dramatically improved luminescence efficiency with an external EL efficiency of 1%.31 These devices are unipolar light-emitting transistors, and their operation mechanism is shown in Figure 7-1. Application of a gate voltage allows holes to accumulate between the electrodes and the organic layer, and an increase in the voltage between the source and drain electrodes allows a region with no hole accumulation (known as a pinch-off region) to be formed near the drain electrode. A strong electric field is created at the interface between the drain electrode and the pinch-off region, accelerating the electron injection.
Schematics and operation mechanisms of light-emitting transistors
Figure 7.Schematics and operation mechanisms of light-emitting transistors
In addition, the development of ambipolar organic semiconductors has lead to higher EL efficiency, and some ambipolar devices (Figure 7-2) have been fabricated with polymer organic semiconductors.32,33 Even in a small-molecule system, a clear electroluminescence has been observed (Figure 8) when a single-crystal from a BSB derivative with a simple molecular frame34 is used. Application of an appropriate gate voltage provides electroluminescence between the source and drain electrodes.
Ambipolar FET producing bright electroluminescence
Figure 8.Ambipolar FET producing bright electroluminescence
This ambipolar type architecture also has many advantages over OLEDs and shows promise as a future device structure for organic semiconductor lasers. Transistors using organic substances have been studied since the early 1980s when a pioneering organic transistor was first reported.35 As findings about materials and devices have accumulated rapidly, the use of fused polycyclic aromatic compounds such as tetracene and rubrene (Product No. 554073) have recently achieved an electron field-effect mobility of about 10 cm2/Vs. Electroluminescence from devices using a transistor structure has a high electronic and optical freedom, and are therefore suitable for organic semiconductor lasers, holding promise for future developments.
Materials
Loading
References
1.
Sorokin PP, Lankard JR. 1966. Stimulated Emission Observed from an Organic Dye, Chloro-aluminum Phthalocyanine. IBM J. Res. & Dev.. 10(2):162-163. https://doi.org/10.1147/rd.102.0162
2.
Schäfer FP, Schmidt W, Volze J. 1966. ORGANIC DYE SOLUTION LASER. Appl. Phys. Lett.. 9(8):306-309. https://doi.org/10.1063/1.1754762
3.
Soffer BH, McFarland BB. 1967. CONTINUOUSLY TUNABLE, NARROW?BAND ORGANIC DYE LASERS. Appl. Phys. Lett.. 10(10):266-267. https://doi.org/10.1063/1.1754804
4.
Sorokin PP, Lankard JR, Moruzzi VL, Hammond EC. 1968. Flashlamp?Pumped Organic?Dye Lasers. The Journal of Chemical Physics. 48(10):4726-4741. https://doi.org/10.1063/1.1668055
5.
Sorokin PP, Lankard JR, Moruzzi VL, Lurio A. 1969. FREQUENCY?LOCKING OF ORGANIC DYE LASERS TO ATOMIC RESONANCE LINES. Appl. Phys. Lett.. 15(6):179-181. https://doi.org/10.1063/1.1652957
6.
Chihaya Adachi. 1989. Student Advanced technical paper. Nippon Kogyo Shinbun.
7.
Tessler N, Denton GJ, Friend RH. 1996. Lasing from conjugated-polymer microcavities. Nature. 382(6593):695-697. https://doi.org/10.1038/382695a0
8.
Frolov SV, Ozaki M, Gellermann W, Vardeny ZV, Yoshino K. 1996. Mirrorless Lasing in Conducting Polymer poly(2,5-dioctyloxy- p-phenylenevinylene) Films. Jpn. J. Appl. Phys.. 35(Part 2, No. 10B):L1371-L1373. https://doi.org/10.1143/jjap.35.l1371
9.
Hide F, Diaz-Garcia MA, Schwartz BJ, Andersson MR, Pei Q, Heeger AJ. 1996. Semiconducting Polymers: A New Class of Solid-State Laser Materials. Science. 273(5283):1833-1836. https://doi.org/10.1126/science.273.5283.1833
10.
Kozlov VG, Bulovi? V, Burrows PE, Forrest SR. 1997. Laser action in organic semiconductor waveguide and double-heterostructure devices. Nature. 389(6649):362-364. https://doi.org/10.1038/38693
11.
Berggren M, Dodabalapur A, Slusher RE, Bao Z. 1997. Light amplification in organic thin films using cascade energy transfer. Nature. 389(6650):466-469. https://doi.org/10.1038/38979
12.
Tessler N. 1999. Lasers Based on Semiconducting Organic Materials. Adv. Mater. 11(5):363-370.
13.
Yamamoto H, Oyamada T, Sasabe H, Adachi C. 2004. Amplified spontaneous emission under optical pumping from an organic semiconductor laser structure equipped with transparent carrier injection electrodes. Appl. Phys. Lett.. 84(8):1401-1403. https://doi.org/10.1063/1.1646730
14.
Adachi C, Baldo MA, Thompson ME, Forrest SR. 2001. Nearly 100% internal phosphorescence efficiency in an organic light-emitting device. Journal of Applied Physics. 90(10):5048-5051. https://doi.org/10.1063/1.1409582
15.
Aimono T, Kawamura Y, Goushi K, Yamamoto H, Sasabe H, Adachi C. 2005. 100% fluorescence efficiency of 4,4[sup ?]-bis[(N-carbazole)styryl]biphenyl in a solid film and the very low amplified spontaneous emission threshold. Appl. Phys. Lett.. 86(7):071110. https://doi.org/10.1063/1.1867555
16.
Nakanotani H, Sasabe H, Adachi C. 2005. Low lasing threshold in organic distributed feedback solid state lasers using bisstyrylbenzene derivative as active material. https://doi.org/10.1117/12.629383
17.
Yamamoto H, Kasajima H, Yokoyama W, Sasabe H, Adachi C. 2005. Extremely-high-density carrier injection and transport over 12000A?cm2 into organic thin films. Appl. Phys. Lett.. 86(8):083502. https://doi.org/10.1063/1.1866230
18.
Nakanotani H, Oyamada T, Kawamura Y, Sasabe H, Adachi C. Injection and Transport of High Current Density over 1000 A/cm2in Organic Light Emitting Diodes under Pulse Excitation. Jpn. J. Appl. Phys.. 44(6A):3659-3662. https://doi.org/10.1143/jjap.44.3659
19.
Baldo MA, Adachi C, Forrest SR. Transient analysis of organic electrophosphorescence. II.?Transient analysis of triplet-triplet annihilation. Phys. Rev. B. 62(16):10967-10977. https://doi.org/10.1103/physrevb.62.10967
20.
Baldo MA, Holmes RJ, Forrest SR. Prospects for electrically pumped organic lasers. Phys. Rev. B. 66(3): https://doi.org/10.1103/physrevb.66.035321
21.
Nakanotani H, Sasabe H, Adachi C. 2005. Singlet-singlet and singlet-heat annihilations in fluorescence-based organic light-emitting diodes under steady-state high current density. Appl. Phys. Lett.. 86(21):213506. https://doi.org/10.1063/1.1939075
22.
Adachi C, Baldo MA, Thompson ME, Forrest SR. 2001. Nearly 100% internal phosphorescence efficiency in an organic light-emitting device. Journal of Applied Physics. 90(10):5048-5051. https://doi.org/10.1063/1.1409582
23.
Yokoyama W, Sasabe H, Adachi C. 2003. Carrier Injection and Transport of Steady-State High Current Density Exceeding 1000 A/cm2in Organic Thin Films*. Jpn. J. Appl. Phys.. 42(Part 2, No. 11B):L1353-L1355. https://doi.org/10.1143/jjap.42.l1353
24.
Holzer W, Penzkofer A, Schmitt T, Hartmann A, Bader C, Tillmann H, Raabe D, Stockmann R, Hörhold H. 2001. 33(2):121-150. https://doi.org/10.1023/a:1007020731786
25.
Liu X, Py C, Tao Y, Li Y, Ding J, Day M. 2004. Low-threshold amplified spontaneous emission and laser emission in a polyfluorene derivative. Appl. Phys. Lett.. 84(15):2727-2729. https://doi.org/10.1063/1.1699479
26.
Shaklee KL, Leheny RF. 1971. DIRECT DETERMINATION OF OPTICAL GAIN IN SEMICONDUCTOR CRYSTALS. Appl. Phys. Lett.. 18(11):475-477. https://doi.org/10.1063/1.1653501
27.
Schön JH. 2000. A Superconducting Field-Effect Switch. 288(5466):656-658. https://doi.org/10.1126/science.288.5466.656
28.
Hepp A, Heil H, Weise W, Ahles M, Schmechel R, von Seggern H. Light-Emitting Field-Effect Transistor Based on a Tetracene Thin Film. Phys. Rev. Lett.. 91(15): https://doi.org/10.1103/physrevlett.91.157406
29.
Sakanoue T, Fujiwara E, Yamada R, Tada H. 2004. Visible light emission from polymer-based field-effect transistors. Appl. Phys. Lett.. 84(16):3037-3039. https://doi.org/10.1063/1.1710713
30.
Oyamada T, Sasabe H, Adachi C, Okuyama S, Shimoji N, Matsushige K. 2005. Electroluminescence of 2,4-bis(4-(2?-thiophene-yl)phenyl)thiophene in organic light-emitting field-effect transistors. Appl. Phys. Lett.. 86(9):093505. https://doi.org/10.1063/1.1870105
31.
Oyamada T, Uchiuzou H, Akiyama S, Oku Y, Shimoji N, Matsushige K, Sasabe H, Adachi C. 2005. Lateral organic light-emitting diode with field-effect transistor characteristics. Journal of Applied Physics. 98(7):074506. https://doi.org/10.1063/1.2060932
32.
Oyamada T, Uchiuzou H, Akiyama S, Oku Y, Shimoji N, Matsushige K, Sasabe H, Adachi C. 2005. Lateral organic light-emitting diode with field-effect transistor characteristics. Journal of Applied Physics. 98(7):074506. https://doi.org/10.1063/1.2060932
33.
Zaumseil J, Friend RH, Sirringhaus H. 2006. Spatial control of the recombination zone in an ambipolar light-emitting organic transistor. Nature Mater. 5(1):69-74. https://doi.org/10.1038/nmat1537
34.
Dinelli F, Capelli R, Loi M, Murgia M, Muccini M, Facchetti A, Marks T. 2006. High-Mobility Ambipolar Transport in Organic Light-Emitting Transistors. Adv. Mater.. 18(11):1416-1420. https://doi.org/10.1002/adma.200502164
35.
Nakanotani H, Saito M, Nakamura H, Adachi C. 2009. Highly balanced ambipolar mobilities with intense electroluminescence in field-effect transistors based on organic single crystal oligo(p-phenylenevinylene) derivatives. Appl. Phys. Lett.. 95(3):033308. https://doi.org/10.1063/1.3184588
36.
Kudo K, Yamashina M, Moriizumi T. 1984. Field Effect Measurement of Organic Dye Films. Jpn. J. Appl. Phys.. 23(Part 1, No. 1):130-130. https://doi.org/10.1143/jjap.23.130